Learn More
Sino Biological Recombinant Rat IL-20R beta/IL-20RB Protein (hFc Tag)
Supplier:  Sino Biological 80187R02H100
Encompass_Preferred
A DNA sequence encoding the rat IL20RB (B8K1X9) (Met1-Pro227) was expressed, fused with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-162-3823