Learn More
Sino Biological Recombinant Rat IL-20R beta/IL-20RB Protein (His Tag)
Supplier: Sino Biological 80187R08H100
A DNA sequence encoding the rat IL20RB (B8K1X9) (Met1-Pro227) was expressed, fused with a polyhistidine tag at the C-terminus.