Learn More
Sino Biological Recombinant Rat IL-21 Receptor/IL-21R Protein (His Tag)
Supplier:  Sino Biological 80189R08H100
Encompass_Preferred
A DNA sequence encoding the rat IL21R (Q5EBB1) (Met1-Pro236) was expressed, fused with a polyhistidine tag at the C-terminus.
Catalog No. 50-162-3829