Learn More
Sino Biological Recombinant Rat IL-17RA Protein (hFc Tag)
Supplier:  Sino Biological 80190R02H100
Encompass_Preferred
A DNA sequence encoding the rat IL17RA (NP_001101353.2)(Met1-Pro320) was expressed with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-162-3831