Learn More
Sino Biological Recombinant Rat IL-17RA Protein (His Tag)
Supplier:  Sino Biological 80190R08H100
Encompass_Preferred
A DNA sequence encoding the rat IL17RA (NP_001101353.2) (Met1-Pro320) was expressed, fused with a polyhistidine tag at the C-terminus.
Catalog No. 50-162-3833