Learn More
Sino Biological Recombinant Rat IL-18Rα Protein (hFc Tag)
Supplier:  Sino Biological 80191R02H100
Encompass_Preferred
A DNA sequence encoding the rat IL18R1 (NP_001100375.1) extracellular domain (Met 1-Gly 326) was fused with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-162-3835