Learn More
Sino Biological Recombinant Rat IL-18Rα Protein (His Tag)
Supplier:  Sino Biological 80191R08H100
Encompass_Preferred
A DNA sequence encoding the rat IL18R1 (D3ZRM2) (Met1-Gly326) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-162-3837