Learn More
Sino Biological Recombinant Rat GITR/TNFRSF18/AITR Protein (hFc Tag)
Supplier:  Sino Biological 80192R02H100
Encompass_Preferred
A DNA sequence encoding the rat TNFRSF18 (Q5M835) (Met1-Lys121) was expressed, fused with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-162-3839