Learn More
Sino Biological Recombinant Rat IL-25/IL-17E Protein (hFc Tag)
Supplier:  Sino Biological 80194R01H50
Encompass_Preferred
A DNA sequence encoding the rat IL25 (D3ZLB1) (Val17-Ala169) was expressed, fused with Fc region of human IgG1 at the N-terminus.
Catalog No. 50-162-3842