Learn More
Sino Biological Recombinant Rat IL1R2 Protein (His Tag)
Supplier:  Sino Biological 80196R08H100
Encompass_Preferred
A DNA sequence encoding the rat IL1R2 (P43303) extracellular domain (Met 1-Glu 355) was expressed, fused with a polyhistidine tag at the C-terminus.
Catalog No. 50-162-3843