Learn More
Sino Biological Recombinant Rat IL-2R gamma/CD132 Protein (hFc Tag)
Supplier:  Sino Biological 80197R02H100
Encompass_Preferred
A DNA sequence encoding the rat IL2RG (Q68FU6) (Met1-Ala262) was expressed, fused with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-162-3845