Learn More
Sino Biological Recombinant Rat IL-2R gamma/CD132 Protein (His Tag)
Supplier:  Sino Biological 80197R08H100
Encompass_Preferred
A DNA sequence encoding the rat IL2RG (Q68FU6) (Met1-Ala262) was expressed, fused with a polyhistidine tag at the C-terminus.
Catalog No. 50-162-3847