Learn More
Sino Biological Recombinant Rat IL-4R Protein (His Tag), HPLC-verified
Supplier:  Sino Biological 80198R08H50
Encompass_Preferred
A DNA sequence encoding the rat IL4R (Q63257-1) (Met1-Arg232) was expressed, fused with a polyhistidine tag at the C-terminus.
Catalog No. 50-162-3852