Learn More
Sino Biological Recombinant Rat IL-21 Protein (His Tag)
Supplier:  Sino Biological 80200R08B100
Encompass_Preferred
A DNA sequence encoding the rat IL21 (NP_001102413.1) (Met1-Ser146) was expressed with a polyhistide tag at the C-terminus.
Catalog No. 50-162-3853