Learn More
Sino Biological Recombinant Rat IL-13 R alpha 2/IL13RA2 Protein (hFc Tag)
Supplier:  Sino Biological 80204R02H100
Encompass_Preferred
A DNA sequence encoding the rat IL13RA2 (Q8VHK6) (Met1-Lys336) was expressed, fused with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-162-3856