Learn More
Sino Biological Recombinant Rat IL-13 R alpha 2/IL13RA2 Protein (His Tag), HPLC-verified
Supplier:  Sino Biological 80204R08H100
Encompass_Preferred
A DNA sequence encoding the rat IL13RA2 (Q8VHK6) (Met1-Lys336) was expressed, fused with a polyhistidine tag at the C-terminus.
Catalog No. 50-162-3858