Learn More
Sino Biological Recombinant Rat interferon-alpha/IFNA5 Protein (His Tag)
Supplier: Sino Biological 80205R08H100
A DNA sequence encoding the rat IFNA5 (XP_001076062.1) (Met1-Glu189) was expressed, fused with a polyhistidine tag at the C-terminus.