Learn More
Sino Biological Recombinant Rat EDA2R/TNFRSF27 Protein (His Tag)
Supplier:  Sino Biological 80206R08H100
Encompass_Preferred
A DNA sequence encoding the rat EDA2R (D3ZAX4) (Met2-Glu137) was expressed, fused with a polyhistidine tag at the C-terminus.
Catalog No. 50-162-3867