Learn More
Sino Biological Recombinant Rat CLEC-1 Protein (hFc Tag)
Supplier:  Sino Biological 80212R01H50
Encompass_Preferred
A DNA sequence encoding the rat CLEC1A (D3ZGU3) (Gln73-Gln269) was expressed, fused with Fc region of human IgG1 at the N-terminus.
Catalog No. 50-162-3876