Learn More
Sino Biological Recombinant Rat CLEC4D Protein (hFc Tag)
Supplier:  Sino Biological 80213R01H100
Encompass_Preferred
A DNA sequence encoding the rat CLEC4D (Q69FH1) (Trp48-Lys218) was expressed with Fc region of human IgG1 at the N-terminus.
Catalog No. 50-162-3877