Learn More
Sino Biological Recombinant Rat CLEC4D Protein (His Tag)
Supplier:  Sino Biological 80213R07H100
Encompass_Preferred
A DNA sequence encoding the rat CLEC4D (Q69FH1) (Trp48-Lys218) was expressed, fused with a polyhistidine tag at the N-terminus.
Catalog No. 50-162-3879