Learn More
Sino Biological Recombinant Rat CLEC4F Protein
Supplier:  Sino Biological 80215RNCH50
Encompass_Preferred
A DNA sequence encoding the rat CLEC4F(NP_446205.1) (Arg70-Ser550) was expressed with two additional amino acids (Gly & Pro) at the N-terminus.
Catalog No. 50-162-3882