Learn More
Sino Biological Recombinant Rat ASAM Protein (His Tag)
Supplier:  Sino Biological 80219R08H100
Encompass_Preferred
A DNA sequence encoding the rat ASAM (Q8K1G0) (Met1-Met232) was expressed, fused with a polyhistidine tag at the C-terminus.
Catalog No. 50-162-3883