Learn More
Sino Biological Recombinant Rat CADM3 Protein (His Tag)
Supplier:  Sino Biological 80220R08H100
Encompass_Preferred
A DNA sequence encoding the rat CADM3 (Q1WIM3) extracellular domain (Met 1-His 328) was expressed, fused with a polyhistidine tag at the C-terminus.
Catalog No. 50-162-3887