Learn More
Sino Biological Recombinant Rat BCAM Protein (His Tag), HPLC-verified
Supplier:  Sino Biological 80222R08H100
Encompass_Preferred
A DNA sequence encoding the rat BCAM (Q9ESS6) (Met1-Ala543) was expressed, fused with a polyhistidine tag at the C-terminus.
Catalog No. 50-162-3893