Learn More
Sino Biological Recombinant Rat CD27/TNFRSF7 Protein (hFc Tag)
Supplier:  Sino Biological 80223R02H100
Encompass_Preferred
A DNA sequence encoding the rat CD27 (NP_001019506.1) (Met1-Arg183) was expressed with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-162-3895