Learn More
Sino Biological Recombinant Rat CD27/TNFRSF7 Protein (mFc Tag)
Supplier:  Sino Biological 80223R38H100
Encompass_Preferred
A DNA sequence encoding the rat CD27 (NP_001019506.1) (Met1-Cys181) was expressed with the Fc region of mouse IgG2a at the C-terminus.
Catalog No. 50-162-3897