Learn More
Sino Biological Recombinant Rat CEACAM-1/CD66a Protein (His Tag)
Supplier:  Sino Biological 80224R08H100
Encompass_Preferred
A DNA sequence encoding the rat CEACAM1 (NP_001029032.1) (Met1-Ser422) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-162-3899