Learn More
Sino Biological Recombinant Rat KIRREL3 Protein (His Tag)
Supplier:  Sino Biological 80225R08H100
Encompass_Preferred
A DNA sequence encoding the rat KIRREL3 (Q09GS6) (Met1-Ala523) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-162-3901