Learn More
Sino Biological Recombinant Rat LAMP1 Protein (hFc Tag)
Supplier:  Sino Biological 80226R02H100
Encompass_Preferred
A DNA sequence encoding the Rat LAMP1 (P14562) (Met1-Asn371) was expressed, fused with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-162-3903