Learn More
Sino Biological Recombinant Rat EDAR Protein (hFc Tag)
Supplier:  Sino Biological 80228R02H100
Encompass_Preferred
A DNA sequence encoding the rat EDAR (NP_001178828.1) extracellular domain (Met 1-Ala 187) was fused with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-162-3905