Learn More
Sino Biological Recombinant Rat Galectin-8/LGALS8 Protein (GST Tag)
Supplier:  Sino Biological 80231R09E100
Encompass_Preferred
A DNA sequence encoding rat LGALS8 (NP_446314.2) (Met 1-Trp 316) was fused with the GST tag at the N-terminus.
Catalog No. 50-162-3911