Learn More
Sino Biological Recombinant Rat MDH1 Protein (His Tag)
Supplier:  Sino Biological 80232R08E50
Encompass_Preferred
A DNA sequence encoding the rat MDH1 (O88989) (Met 4-Ala 334) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-162-3914