Learn More
Sino Biological Recombinant Rat IFN gamma Protein (hFc Tag)
Supplier:  Sino Biological 80234R02H100
Encompass_Preferred
A DNA sequence encoding the rat IFNG (P01581) (Met 1-Cys 156) was fused with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-162-3915