Learn More
Sino Biological Recombinant Rat JAM-A/F11 Receptor Protein (hFc Tag)
Supplier:  Sino Biological 80240R02H100
Encompass_Preferred
A DNA sequence encoding the rat F11R () was expressed, fused with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-162-3919