Learn More
Sino Biological Recombinant Rat JAM-A/F11 Receptor Protein (His Tag)
Supplier:  Sino Biological 80240R08H50
Encompass_Preferred
A DNA sequence encoding the rat F11R (Q9JHY1) (Met1-Gly238) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-162-3922