Learn More
Sino Biological Recombinant Rat ESAM Protein (hFc Tag)
Supplier:  Sino Biological 80241R02H100
Encompass_Preferred
A DNA sequence encoding the rat ESAM (Q6AYD4) extracellular domain (Met 1-Ala 251) was fused with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-162-3923