Learn More
Sino Biological Recombinant Rat HEPACAM2 Protein (hFc Tag)
Supplier:  Sino Biological 80242R02H100
Encompass_Preferred
A DNA sequence encoding the rat HepaCAM2 (B5DEN8) (Met1-Leu350) was expressed with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-162-3925