Learn More
Sino Biological Recombinant Rat TrkB Protein (hFc Tag)
Supplier:  Sino Biological 80243R02H20
Encompass_Preferred
A DNA sequence encoding the Rat NTRK2 (NP_036863) (Met1-His429) was expressed, fused with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-162-3929