Learn More
Sino Biological Recombinant Rat TrkB Protein (His Tag)
Supplier:  Sino Biological 80243R08H20
Encompass_Preferred
A DNA sequence encoding the rat NTRK2 (Q63604-Isoform T1) (Met1-His429) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-162-3931