Learn More
Andwin Scientific 81 WELL FREEZER RACK ASSTD 5PK
SDP
Supplier:  Andwin Scientific 61600A
Encompass_Preferred
The polypropylene 81-Well Freezer Storage Rack fits in 5.25 in. square freezer modules and holds 1.5-2ml microtubes and vials. It features imprinted coordinates, molded grid lines and reference points; sold in a pack of 5 of assorted colors.
Catalog No. 50-727-576