Learn More
eMolecules​ 823-78-9 | 3-Bromobenzyl bromide | Combi-Blocks | MFCD00000176 | 249.933 | C7H6Br2 | 98.000 | BrCc1cccc(Br)c1 | 25g | 117539755
Supplier:  eMolecules​ 117539755
Encompass_Preferred
3-Bromobenzyl bromide | Combi-Blocks | 823-78-9 | MFCD00000176 | 249.933 | C7H6Br2 | 98.000 | BrCc1cccc(Br)c1 | 25g | 117539755
Catalog No. 50-004-92308