Learn More
eMolecules​ 824-54-4 | 2-Bromo-4-methylbenzaldehyde | Ambeed | MFCD04039890 | 199.047 | C8H7BrO | 97.000 | Cc1ccc(C=O)c(Br)c1 | 500g | 534084087
Supplier:  eMolecules​ 534084087
Encompass_Preferred
2-Bromo-4-methylbenzaldehyde | Ambeed | 824-54-4 | MFCD04039890 | 199.047 | C8H7BrO | 97.000 | Cc1ccc(C=O)c(Br)c1 | 500g | 534084087
Catalog No. 50-001-59272