Learn More
eMolecules​ 847818-74-0 | 1-Methyl-1H-pyrazole-5-boronic acid pinacol ester | Combi-Blocks | MFCD05861380 | 208.070 | C10H17BN2O2 | 98.000 | Cn1nccc1B1OC(C)(C)C(C)(C)O1 | 100g | 267168061
Supplier:  eMolecules​ 267168061
Encompass_Preferred
1-Methyl-1H-pyrazole-5-boronic acid pinacol ester | Combi-Blocks | 847818-74-0 | MFCD05861380 | 208.070 | C10H17BN2O2 | 98.000 | Cn1nccc1B1OC(C)(C)C(C)(C)O1 | 100g | 267168061
Catalog No. 50-004-91892