Learn More
CPC Scientific H-Thr-Tyr-Ile-Cys(Bzl)-Glu(OBzl)-Val-Glu-Asp-Gln-Lys-Glu-Glu-OH (ammonium salt) 25MG
SDP
Supplier:  CPC Scientific HIV005B
Encompass
This dibenzylated derivative was found to completely inhibit HIV-1-induced cell fusion and infection in vitroONE-LETTER SEQUENCE: TYIC(Bzl)-E(OBzl)-VEDQKEEMOLECULAR FORMULA: C76H108N14O26S1MOLECULAR WEIGHT:1665.84STORAGE CONDITIONS: -20 5CRESEARCH AREA: AntimicrobialREFERENCES: J.D.Lifson et al., Science, 241, 712 (1988); P.L.Nara et al., Proc. Natl. Acad. Sci. USA, 86, 7139 (1989); H.Repke et al., J. Immunol., 149, 1809 (1992)
Catalog No. 50-210-9538