Learn More
Bellco Glass, Inc. 850mL Suspension Culture
SDP
Supplier:  Bellco Glass, Inc. 229540000
Encompass_Preferred
Flask is printed with lot number and expiration date for easy identification and tracking. Molded interlocking T design for improved stacking on 600 amp850mL size flasks. Vent caps feature 0.22µm hydrophobic membrane.Specifications Convenient zipclosure bag is a practical alternative to folding and taping open bags Large opening canted neck design and moderate angles allow easy access to entire flask surface ideal for cell scrapers Stable base enables filling with flask standing upright Thick durable walls Size 850mL Pack Description 3/Resealable Bag Case Quantity 18
Catalog No. 50-232-9943