Learn More
CTMATTER LLC 8540016770076, SKILCRAFT Multi-Fold Paper Towel, 1-Ply, 9.25 x 3, White, 250/Bundle, 16 Bundles/Box
Supplier:  CTMATTER LLC NSN6770076
Encompass_Preferred
Save money by replacing C-fold paper towels, which can over-dispense, with these three-panel multi-fold paper towels. Free of processed chlorine. Towels measure 9.25 x 3 inches folded and open to 9.25 x 9.5 inches.
Catalog No. NC3861256