Learn More
Refrigiwear Inc. 855L - 0227R Regular Silver L/XL Touch Screen Glove
SDP
Supplier:  Refrigiwear Inc. 0227RSLVLXL
Encompass_Preferred
Gloves don't do you much good if you take them off all the time. The Touchscreen Glove allows use of touchscreens while keeping your fingers cozy and warm! Soft acrylic stretch knit is never itchy, and stretches to fit large or small hands. Silver conductive thread on all five fingers allows you to use all your touchscreen devices, including iPads, iPhones, Android phones, and tablets. Acrylic knit with PVC dot grip on palm Special fingertips allows for use of touchscreens Wear with fingerless mitts for extra warmth and added touchscreen functionality  
Catalog No. NC3867066