Learn More
Labrepco Inc -86C ULT CHST FRZR DUAL COOL
SDP
Supplier:  Labrepco Inc MDFDC700VXCPA
Encompass_Preferred
PHCbi TwinGuard® Series 25.3 Cu. Ft. (Capacity: 507 x 2"Boxes) -86°C ULT Chest Freezer with Dual Cool RefrigerationTechnology (208/230V)
Catalog No. NC2588495
Only null left
Add to Cart
Request a Quote