Learn More
Cayman Chemical 8bromo CyclIc AMPsodIM sa 10mg
SDP
Supplier:  Cayman Chemical 1443110 MG
Encompass_Preferred
A PKA activator (Ka = 0.05 µM); selective for PKA over PKG (Ka = 5.8 µM); more resistant to hydrolysis by PDEs than cAMP; inhibits the proliferation of HL-60 cells (IC50 = 18 µM after six days); induces relaxation of phenylephrine-precontracted isolated rabbit carotid or renal arterial rings at 10-300 µM; intradermal administration prevents SLIGRL-NH2-induced scratching and increases in cutaneous LTB4 production in mice at 10 nmol/site
Catalog No. 50-297-1082